This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Gg03323172_m1
See other DISP3 GE Assays ›
Gene Symbol:
DISP3
Gene Name:
dispatched RND transporter family member 3
Gene Aliases:
PTCHD2, TRUP1
Chromosome Location:
Chr.21: 5717723 - 5834841 on Build GRCg6a
Species:
Chicken
Species Specific ID (Flybase ID):
-
Assay ID Gg03323172_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: DISP3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001199176.1 NP_001186105.1 2 - 3 1153 73
XM_015296920.2 XP_015152406.1 3 - 4 1565 73
XM_015296921.2 XP_015152407.1 3 - 4 1568 73
XM_015296922.2 XP_015152408.1 3 - 4 1268 73
XM_015296924.2 XP_015152410.1 3 - 4 1395 73
XM_015296925.2 XP_015152411.1 9 - 10 6922 73
XM_015296926.2 XP_015152412.1 3 - 4 2027 73
XM_015296927.2 XP_015152413.1 3 - 4 1665 73
XM_015296928.2 XP_015152414.1 3 - 4 4437 73
GenBank mRNA CR386085.1 - 2 - 3 251 73
EU429800.1 - 2 - 3 1153 73

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Recommended Assay Gallus gallus Probe spans exons

Gene Ontology Categories:

Function(s)

cholesterol metabolic process
response to hormone
cholesterol homeostasis
negative regulation of neuron differentiation
positive regulation of lipid metabolic process
positive regulation of neural precursor cell proliferation

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0