This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Gg03324428_m1
See other MSANTD4 GE Assays ›
Gene Symbol:
MSANTD4
Gene Name:
Myb/SANT DNA binding domain containing 4 with coiled-coils
Gene Aliases:
-
Chromosome Location:
Chr.1: 182456211 - 182471903 on Build GRCg6a
Species:
Chicken
Species Specific ID (Flybase ID):
-
Assay ID Gg03324428_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MSANTD4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_004938836.3 XP_004938893.1 2 - 3 1183 81
XM_015279651.2 XP_015135137.1 1 - 2 2536 81
XM_015279668.2 XP_015135154.1 2 - 3 673 81
XM_015279670.2 XP_015135156.1 2 - 3 2948 81
XM_417171.6 XP_417171.2 2 - 3 1643 81
GenBank mRNA CR388777.1 - 1 - 2 271 81

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Recommended Assay Gallus gallus Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0