This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Gg03325162_m1
See other SCLY GE Assays ›
Gene Symbol:
SCLY
Gene Name:
selenocysteine lyase
Gene Aliases:
-
Chromosome Location:
Chr.9: 896101 - 904990 on Build GRCg6a
Species:
Chicken
Species Specific ID (Flybase ID):
-
Assay ID Gg03325162_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SCLY
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001139463.1 NP_001132935.1 10 - 11 1196 54
NM_001271525.2 NP_001258454.1 11 - 12 1187 54
XM_015291343.2 XP_015146829.1 11 - 12 1466 54
XM_015291344.2 XP_015146830.1 11 - 12 1464 54
XM_015291345.2 XP_015146831.1 11 - 12 1466 54
XM_015291346.2 XP_015146832.1 11 - 12 1466 54
XM_015291347.2 XP_015146833.1 11 - 12 1244 54
XM_015291348.2 XP_015146834.1 11 - 12 1264 54
XM_025153330.1 XP_025009098.1 11 - 12 1159 54
GenBank mRNA BU398435.1 - 2 - 3 158 54
CR406321.1 - 10 - 11 988 54

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Recommended Assay Gallus gallus Probe spans exons

Panther Classification:

Molecular Function -

lyase metabolite interconversion enzyme

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-vnb2r:80/100.66.79.197:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0