This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Gg03326253_m1
See other BPHL GE Assays ›
Gene Symbol:
BPHL
Gene Name:
biphenyl hydrolase like
Gene Aliases:
-
Chromosome Location:
Chr.2: 66284215 - 66312335 on Build GRCg6a
Species:
Chicken
Species Specific ID (Flybase ID):
-
Assay ID Gg03326253_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: BPHL
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_015276018.2 XP_015131504.1 1 - 2 125 77
XM_015276019.2 XP_015131505.1 1 - 2 125 77
XM_025147673.1 XP_025003441.1 1 - 2 125 77
XM_025147674.1 XP_025003442.1 1 - 2 125 77
XM_025147675.1 XP_025003443.1 1 - 2 125 77
GenBank mRNA BX930605.2 - 1 - 2 88 77
BX932337.1 - 1 - 2 98 77

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Recommended Assay Gallus gallus Probe spans exons

Panther Classification:

Molecular Function -

hydrolase metabolite interconversion enzyme

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-ng27p:80/100.66.78.145:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline