This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Gg03326651_m1
See other HPGD GE Assays ›
Gene Symbol:
HPGD
Gene Name:
hydroxyprostaglandin dehydrogenase 15-(NAD)
Gene Aliases:
-
Chromosome Location:
Chr.4: 43759685 - 43792292 on Build GRCg6a
Species:
Chicken
Species Specific ID (Flybase ID):
-
Assay ID Gg03326651_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: HPGD
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001277784.1 NP_001264713.1 4 - 5 546 83
XM_015276239.2 XP_015131725.1 2 - 3 219 83
XM_025149634.1 XP_025005402.1 5 - 6 1729 83
GenBank mRNA BU258980.1 - 4 - 5 554 83
BX929847.1 - 4 - 5 554 83
BX934937.1 - 4 - 5 494 83
CR353827.1 - 4 - 5 468 83

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Recommended Assay Gallus gallus Probe spans exons

Panther Classification:

Molecular Function -

dehydrogenase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0