This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Gg03327495_m1
See other KMT5B GE Assays ›
Gene Symbol:
KMT5B
Gene Name:
lysine methyltransferase 5B
Gene Aliases:
CGI-85, SUV420H1
Chromosome Location:
Chr.5: 16368467 - 16422509 on Build GRCg6a
Species:
Chicken
Species Specific ID (Flybase ID):
-
Assay ID Gg03327495_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: KMT5B
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_015286821.2 XP_015142307.1 6 - 7 1656 70
XM_015286822.2 XP_015142308.1 5 - 6 650 70
XM_015286823.2 XP_015142309.1 4 - 5 583 70
GenBank mRNA BX929560.2 - 5 - 6 753 70

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Gallus gallus Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0