This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Gg03328879_m1
See other RAC2 GE Assays ›
Gene Symbol:
RAC2
Gene Name:
ras-related C3 botulinum toxin substrate 2 (rho family, small GTP binding protein Rac2)
Gene Aliases:
-
Chromosome Location:
Chr.1: 51413925 - 51423026 on Build GRCg6a
Species:
Chicken
Species Specific ID (Flybase ID):
-
Assay ID Gg03328879_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: RAC2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001201452.1 NP_001188381.1 5 - 6 624 103
GenBank mRNA AJ398007.1 - 5 - 6 435 103
AJ851813.1 - 5 - 6 588 103
CF074925.1 - 5 - 6 570 103
CR353911.1 - 5 - 6 624 103

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Recommended Assay Gallus gallus Probe spans exons

Panther Classification:

Molecular Function -

small GTPase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-bm6jx:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0