Assay Details
| Gene Symbol: SWI5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AY662689.1 | - | 1 - 2 | 47 | 87 | |
| CR523161.1 | - | 2 - 3 | 253 | 87 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SWI5| Gene Symbol: SWI5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AY662689.1 | - | 1 - 2 | 47 | 87 | |
| CR523161.1 | - | 2 - 3 | 253 | 87 | ||
Set Membership: |
|