This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Gg03337959_g1
See other CISH GE Assays ›
Gene Symbol:
CISH
Gene Name:
cytokine inducible SH2 containing protein
Gene Aliases:
CIS
Chromosome Location:
Chr.12: 2264031 - 2268470 on Build GRCg6a
Species:
Chicken
Species Specific ID (Flybase ID):
-
Assay ID Gg03337959_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer:
435.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CISH
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_204626.1 NP_989957.1 2 - 3 477 100
GenBank mRNA AF167297.1 - 2 - 3 477 100

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Gallus gallus Probe spans exons

Panther Classification:

Molecular Function -

kinase modulator

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of signal transduction
protein ubiquitination
intracellular signal transduction
regulation of growth
regulation of phosphatidylinositol 3-kinase activity
phosphatidylinositol phosphorylation
1-phosphatidylinositol-3-kinase regulator activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-49tpj:80/100.66.77.222:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline