This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Gg03338118_m1
See other EIF2AK1 GE Assays ›
Gene Symbol:
EIF2AK1
Gene Name:
eukaryotic translation initiation factor 2 alpha kinase 1
Gene Aliases:
-
Chromosome Location:
Chr.14: 1362506 - 1379879 on Build GRCg6a
Species:
Chicken
Species Specific ID (Flybase ID):
-
Assay ID Gg03338118_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: EIF2AK1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_204648.1 NP_989979.1 3 - 4 698 65
XM_015294130.2 XP_015149616.1 3 - 4 523 65
XM_015294131.2 XP_015149617.1 2 - 3 281 65
XM_025155044.1 XP_025010812.1 4 - 5 600 65
XM_025155045.1 XP_025010813.1 4 - 5 613 65
XM_025155046.1 XP_025010814.1 4 - 5 625 65
XM_025155047.1 XP_025010815.1 3 - 4 523 65
XM_025155048.1 XP_025010816.1 2 - 3 818 65
GenBank mRNA AF330008.1 - 3 - 4 698 65

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Gallus gallus Probe spans exons

Panther Classification:

Molecular Function -

non-receptor serine/threonine protein kinase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8cb8z:80/100.66.72.77:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0