This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Gg03338461_m1
See other OPCML GE Assays ›
Gene Symbol:
OPCML
Gene Name:
opioid binding protein/cell adhesion molecule like
Gene Aliases:
OBCAM
Chromosome Location:
Chr.24: 2142669 - 2396898 on Build GRCg6a
Species:
Chicken
Species Specific ID (Flybase ID):
-
Assay ID Gg03338461_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer:
435.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: OPCML
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_204687.2 NP_990018.1 2 - 3 418 87
XM_015297949.2 XP_015153435.1 3 - 4 607 87
XM_015297950.2 XP_015153436.1 4 - 5 674 87
XM_015297951.2 XP_015153437.1 3 - 4 1168 87
XM_025143171.1 XP_024998939.1 4 - 5 681 87
XM_025143172.1 XP_024998940.1 3 - 4 1036 87
GenBank mRNA AF292934.1 - 2 - 3 418 87
Y08170.2 - 395 87

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Recommended Assay Gallus gallus Probe spans exons

Panther Classification:

Molecular Function -

immunoglobulin

Gene Ontology Categories:

Function(s)

cell adhesion

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-49tpj:80/100.66.77.222:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline