This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Gg03338819_m1
See other HDAC2 GE Assays ›
Gene Symbol:
HDAC2
Gene Name:
histone deacetylase 2
Gene Aliases:
-
Chromosome Location:
Chr.3: 64830904 - 64853665 on Build GRCg6a
Species:
Chicken
Species Specific ID (Flybase ID):
-
Assay ID Gg03338819_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: HDAC2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_204831.2 NP_990162.2 4 - 5 431 76
XM_015284494.2 XP_015139980.1 5 - 6 657 76
GenBank mRNA AF039752.1 - 4 - 5 431 76

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Recommended Assay Gallus gallus Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

chromatin organization
positive regulation of transcription from RNA polymerase II promoter
histone H3 deacetylation
histone H4 deacetylation
histone deacetylase activity
transcription factor binding
NAD-dependent histone deacetylase activity (H3-K14 specific)

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0