This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Gg03339853_g1
See other PRSS2 GE Assays ›
Gene Symbol:
PRSS2 LOC107054567 LOC107054568
Gene Name:
protease, serine 2
trypsin II-P29
trypsin II-P29-like
Gene Aliases:
-
-
-
Chromosome Location:
Chr.1: 78802144 - 78805436 on Build GRCg6a
Chromosome Location:
Chr.1: 78811808 - 78815352 on Build GRCg6a
Chromosome Location:
Chr.1: 78832612 - 78836410 on Build GRCg6a
Species:
Chicken
Species Specific ID (Flybase ID):
-
-
-
Assay ID Gg03339853_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PRSS2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_205384.1 NP_990715.1 1 - 2 63 79
GenBank mRNA BX931578.2 - 1 - 2 64 79
BX933684.1 - 1 - 2 85 79
CR390911.1 - 1 - 2 64 79
U15157.1 - 1 - 2 63 79
Gene Symbol: LOC107054567
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_015294158.2 XP_015149644.1 1 - 2 160 79
Gene Symbol: LOC107054568
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_015294165.2 XP_015149651.1 1 - 2 78 79

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Detect Genomic DNA Gallus gallus Probe spans exons

Panther Classification:

Molecular Function -

serine protease

Gene Ontology Categories:

Function(s) Process(es)

proteolysis
digestion
serine-type endopeptidase activity
metal ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-jbr9k:80/100.66.74.50:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0