This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Gg03341281_m1
See other CCT5 GE Assays ›
Gene Symbol:
CCT5
Gene Name:
chaperonin containing TCP1 subunit 5
Gene Aliases:
-
Chromosome Location:
Chr.2: 78405520 - 78413505 on Build GRCg6a
Species:
Chicken
Species Specific ID (Flybase ID):
-
Assay ID Gg03341281_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CCT5
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001012563.2 NP_001012581.1 3 - 4 369 86
GenBank mRNA AJ851489.1 - 3 - 4 370 86

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Recommended Assay Gallus gallus Probe spans exons

Panther Classification:

Molecular Function -

chaperonin

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-49tpj:80/100.66.77.222:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline