This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Gg03342089_m1
See other HAGH GE Assays ›
Gene Symbol:
HAGH
Gene Name:
hydroxyacylglutathione hydrolase
Gene Aliases:
-
Chromosome Location:
Chr.14: 6439891 - 6449593 on Build GRCg6a
Species:
Chicken
Species Specific ID (Flybase ID):
-
Assay ID Gg03342089_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: HAGH
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001012789.2 NP_001012807.1 7 - 8 895 60
XM_015294193.1 XP_015149679.1 7 - 8 811 60
XM_015294194.2 XP_015149680.1 8 - 9 766 60
XM_015294195.1 XP_015149681.1 7 - 8 890 60
XM_015294196.2 XP_015149682.1 8 - 9 773 60
XM_015294197.2 XP_015149683.1 9 - 10 927 60
XM_015294198.2 XP_015149684.1 9 - 10 838 60
XM_015294199.1 XP_015149685.1 9 - 10 833 60
XM_015294200.2 XP_015149686.1 7 - 8 865 60
XM_015294201.2 XP_015149687.1 7 - 8 745 60
XM_025155091.1 XP_025010859.1 8 - 9 780 60
XM_025155092.1 XP_025010860.1 8 - 9 788 60
XM_025155093.1 XP_025010861.1 10 - 11 861 60
XM_025155094.1 XP_025010862.1 10 - 11 902 60
XM_025155095.1 XP_025010863.1 8 - 9 887 60
XM_025155096.1 XP_025010864.1 7 - 8 766 60
GenBank mRNA AJ720961.1 - 7 - 8 896 60

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Gallus gallus Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

methylglyoxal catabolic process to D-lactate via S-lactoyl-glutathione
L-ascorbic acid biosynthetic process
hydroxyacylglutathione hydrolase activity
metal ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zpsr4:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0