This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Gg03342851_m1
See other RBM24 GE Assays ›
Gene Symbol:
RBM24
Gene Name:
RNA binding motif protein 24
Gene Aliases:
-
Chromosome Location:
Chr.2: 61921863 - 61947358 on Build GRCg6a
Species:
Chicken
Species Specific ID (Flybase ID):
-
Assay ID Gg03342851_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: RBM24
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001012863.3 NP_001012881.2 3 - 4 697 89
GenBank mRNA AJ719481.1 - 3 - 4 694 89
BX933592.2 - 3 - 4 672 89
CR385490.1 - 3 - 4 467 89

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 3' Most Gallus gallus Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

regulation of alternative mRNA splicing, via spliceosome
mRNA processing
cellular response to DNA damage stimulus
RNA splicing
regulation of myotube differentiation
positive regulation of myotube differentiation
cell differentiation
regulation of mRNA stability
positive regulation of myoblast differentiation
mRNA stabilization
mRNA destabilization
3'-UTR-mediated mRNA destabilization
positive regulation of skeletal muscle fiber differentiation
positive regulation of 3'-UTR-mediated mRNA stabilization
positive regulation of stem cell differentiation
negative regulation of cytoplasmic translation
mRNA 3'-UTR binding
mRNA 3'-UTR AU-rich region binding
pre-mRNA intronic binding
mRNA CDS binding
sequence-specific mRNA binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-nbrrd:80/100.66.77.69:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0