This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Gg03343487_m1
See other GTDC1 GE Assays ›
Gene Symbol:
GTDC1
Gene Name:
glycosyltransferase like domain containing 1
Gene Aliases:
-
Chromosome Location:
Chr.7: 33102828 - 33278422 on Build GRCg6a
Species:
Chicken
Species Specific ID (Flybase ID):
-
Assay ID Gg03343487_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GTDC1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001012921.1 NP_001012939.1 6 - 7 1254 93
XM_015289843.2 XP_015145329.1 7 - 8 1318 93
XM_025152215.1 XP_025007983.1 6 - 7 1085 93
XM_025152217.1 XP_025007985.1 8 - 9 1404 93
XM_025152218.1 XP_025007986.1 6 - 7 1112 93
XM_025152219.1 XP_025007987.1 7 - 8 1274 93
XM_025152220.1 XP_025007988.1 7 - 8 1322 93
XM_025152221.1 XP_025007989.1 7 - 8 1242 93
XM_025152222.1 XP_025007990.1 6 - 7 1187 93
XM_025152223.1 XP_025007991.1 6 - 7 1191 93
XM_025152224.1 XP_025007992.1 5 - 6 1051 93
XM_025152225.1 XP_025007993.1 6 - 7 1156 93
XM_025152226.1 XP_025007994.1 6 - 7 1083 93
XM_025152227.1 XP_025007995.1 6 - 7 1083 93
XM_025152228.1 XP_025007996.1 6 - 7 1083 93
XM_025152229.1 XP_025007997.1 8 - 9 1401 93
XM_025152231.1 XP_025007999.1 7 - 8 1258 93
XM_025152232.1 XP_025008000.1 5 - 6 932 93
XM_025152233.1 XP_025008001.1 6 - 7 1151 93
XM_025152234.1 XP_025008002.1 6 - 7 1156 93
XM_025152235.1 XP_025008003.1 6 - 7 1151 93
XM_025152236.1 XP_025008004.1 6 - 7 1151 93
XM_025152237.1 XP_025008005.1 4 - 5 809 93
XM_025152239.1 XP_025008007.1 7 - 8 1314 93
XM_025152240.1 XP_025008008.1 5 - 6 931 93
XM_025152241.1 XP_025008009.1 9 - 10 1562 93
XM_025152242.1 XP_025008010.1 6 - 7 1107 93
XM_025152243.1 XP_025008011.1 5 - 6 974 93
XM_025152244.1 XP_025008012.1 2 - 3 416 93
GenBank mRNA AJ720944.1 - 6 - 7 1254 93

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 3' Most Recommended Assay Gallus gallus Probe spans exons

Panther Classification:

Molecular Function -

glycosyltransferase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0