This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Gg03343851_m1
See other CEL GE Assays ›
Gene Symbol:
CEL
Gene Name:
carboxyl ester lipase
Gene Aliases:
-
Chromosome Location:
Chr.17: 7073101 - 7078463 on Build GRCg6a
Species:
Chicken
Species Specific ID (Flybase ID):
-
Assay ID Gg03343851_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CEL
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001012997.1 NP_001013015.1 8 - 9 1098 72
GenBank mRNA BX931580.2 - 4 - 5 482 72
BX931585.2 - 8 - 9 1096 72
BX931587.2 - 3 - 4 411 72
BX931605.1 - 3 - 4 339 72
BX933612.1 - 3 - 4 285 72
BX950478.1 - 8 - 9 1098 72
CR386746.1 - 3 - 4 370 72

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Recommended Assay Gallus gallus Probe spans exons

Panther Classification:

Molecular Function -

cell adhesion molecule

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mvcqg:80/100.66.75.199:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0