This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Gg03344370_m1
See other ARFGAP1 GE Assays ›
Gene Symbol:
ARFGAP1
Gene Name:
ADP ribosylation factor GTPase activating protein 1
Gene Aliases:
-
Chromosome Location:
Chr.20: 8785015 - 8802773 on Build GRCg6a
Species:
Chicken
Species Specific ID (Flybase ID):
-
Assay ID Gg03344370_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ARFGAP1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001030857.1 NP_001026028.1 13 - 14 1106 84
XM_025142209.1 XP_024997977.1 14 - 15 1178 84
XM_025142210.1 XP_024997978.1 14 - 15 1175 84
XM_025142211.1 XP_024997979.1 13 - 14 1148 84
XM_025142212.1 XP_024997980.1 13 - 14 1112 84
XM_025142213.1 XP_024997981.1 13 - 14 1109 84
XM_025142214.1 XP_024997982.1 12 - 13 1091 84
XM_025142215.1 XP_024997983.1 12 - 13 1007 84
XM_025142216.1 XP_024997984.1 12 - 13 1082 84
XM_025142217.1 XP_024997985.1 12 - 13 1079 84
XM_025142218.1 XP_024997986.1 12 - 13 1055 84
XM_025142219.1 XP_024997987.1 11 - 12 1031 84
XM_025142220.1 XP_024997988.1 11 - 12 1025 84
XM_025142221.1 XP_024997989.1 13 - 14 1084 84
XM_025142222.1 XP_024997990.1 13 - 14 967 84
GenBank mRNA AJ851570.1 - 13 - 14 1106 84

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Recommended Assay Gallus gallus Probe spans exons

Panther Classification:

Molecular Function -

G-protein modulator protein-binding activity modulator GTPase-activating protein

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2xklz:80/100.66.76.139:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0