This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Gg03344475_m1
See other TNFRSF18 GE Assays ›
Gene Symbol:
TNFRSF18
Gene Name:
TNF receptor superfamily member 18
Gene Aliases:
GITR
Chromosome Location:
Chr.21: 2563494 - 2569804 on Build GRCg6a
Species:
Chicken
Species Specific ID (Flybase ID):
-
Assay ID Gg03344475_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: TNFRSF18
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001030874.1 NP_001026045.1 2 - 3 450 70
GenBank mRNA AJ719323.1 - 2 - 3 450 70
MH492023.1 - 2 - 3 309 70

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Gallus gallus Probe spans exons

Panther Classification:

Molecular Function -

transmembrane signal receptor

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zpsr4:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0