This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Gg03344786_m1
See other SYNCRIP GE Assays ›
Gene Symbol:
SYNCRIP
Gene Name:
synaptotagmin binding cytoplasmic RNA interacting protein
Gene Aliases:
-
Chromosome Location:
Chr.3: 76879334 - 76908036 on Build GRCg6a
Species:
Chicken
Species Specific ID (Flybase ID):
-
Assay ID Gg03344786_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SYNCRIP
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001031083.1 NP_001026254.1 4 - 5 514 113
XM_015284620.2 XP_015140106.2 4 - 5 4169 113
XM_015284621.2 XP_015140107.2 4 - 5 4169 113
XM_015284622.2 XP_015140108.1 4 - 5 515 113
XM_015284623.2 XP_015140109.2 4 - 5 4169 113
XM_015284624.2 XP_015140110.2 4 - 5 515 113
XM_025148759.1 XP_025004527.1 4 - 5 4169 113
XM_025148760.1 XP_025004528.1 4 - 5 4169 113
GenBank mRNA AJ719757.1 - 4 - 5 537 113

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Recommended Assay Gallus gallus Probe spans exons

Panther Classification:

Molecular Function -

RNA metabolism protein

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8t6tc:80/100.66.75.151:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0