Assay Details
| Gene Symbol: SLC16A9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031222.1 | NP_001026393.1 | 6 - 7 | 1599 | 103 | |
| XM_015288131.2 | XP_015143617.2 | 7 - 8 | 1690 | 103 | ||
| XM_015288132.2 | XP_015143618.2 | 6 - 7 | 1485 | 103 | ||
| XM_015288133.2 | XP_015143619.2 | 7 - 8 | 1665 | 103 | ||
| XM_015288134.2 | XP_015143620.2 | 7 - 8 | 1881 | 103 | ||
| XM_015288135.2 | XP_015143621.2 | 6 - 7 | 1763 | 103 | ||
| GenBank mRNA | AJ720344.1 | - | 6 - 7 | 1599 | 103 | |
More Information
Set Membership: |
|

SLC16A9
Genomic Map