This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Gg03345342_m1
See other AUH GE Assays ›
Gene Symbol:
AUH
Gene Name:
AU RNA binding methylglutaconyl-CoA hydratase
Gene Aliases:
-
Chromosome Location:
Chr.Z: 44207761 - 44329045 on Build GRCg6a
Species:
Chicken
Species Specific ID (Flybase ID):
-
Assay ID Gg03345342_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: AUH
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001031429.1 NP_001026600.1 1 - 2 242 75
NM_001252606.1 NP_001239535.1 1 - 2 242 75
XM_015280295.2 XP_015135781.1 1 - 2 268 75
XM_015280296.2 XP_015135782.1 1 - 2 266 75
XM_025144720.1 XP_025000488.1 1 - 2 266 75
XM_025144721.1 XP_025000489.1 1 - 2 277 75
XM_025144722.1 XP_025000490.1 1 - 2 288 75
XM_025144723.1 XP_025000491.1 1 - 2 282 75
GenBank mRNA AJ719546.1 - 1 - 2 242 75
BU317418.1 - 1 - 2 217 75
CR389254.1 - 1 - 2 241 75

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Recommended Assay Gallus gallus Probe spans exons

Panther Classification:

Molecular Function -

hydratase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2xklz:80/100.66.76.139:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0