This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Gg03345851_m1
See other FGF13 GE Assays ›
Gene Symbol:
FGF13
Gene Name:
fibroblast growth factor 13
Gene Aliases:
FHF-2
Chromosome Location:
Chr.4: 4747075 - 4962390 on Build GRCg6a
Species:
Chicken
Species Specific ID (Flybase ID):
-
Assay ID Gg03345851_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: FGF13
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001001743.1 NP_001001743.1 2 - 3 339 81
XM_015278206.2 XP_015133692.1 3 - 4 645 81
XM_015278207.2 XP_015133693.1 2 - 3 745 81
XM_015278209.2 XP_015133695.1 2 - 3 260 81
GenBank mRNA AF108755.1 - 2 - 3 497 81
AF199610.1 - 2 - 3 339 81
AF199611.1 - 2 - 3 613 81

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Recommended Assay Gallus gallus Probe spans exons

Panther Classification:

Molecular Function -

growth factor intercellular signal molecule

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-mnfm6:80/100.66.78.37:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline