This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Gg03347260_m1
See other P2RX5 GE Assays ›
Gene Symbol:
P2RX5
Gene Name:
purinergic receptor P2X 5
Gene Aliases:
-
Chromosome Location:
Chr.19: 6768151 - 6787281 on Build GRCg6a
Species:
Chicken
Species Specific ID (Flybase ID):
-
Assay ID Gg03347260_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: P2RX5
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_204748.1 NP_990079.1 7 - 8 791 77
XM_015295667.2 XP_015151153.1 7 - 8 1554 77
XM_025141892.1 XP_024997660.1 7 - 8 1557 77
XM_025141893.1 XP_024997661.1 7 - 8 842 77
XM_025141894.1 XP_024997662.1 10 - 11 3919 77
XM_025141895.1 XP_024997663.1 10 - 11 4230 77
XM_025141896.1 XP_024997664.1 11 - 12 1625 77
XM_025141897.1 XP_024997665.1 10 - 11 6127 77
XM_025141898.1 XP_024997666.1 10 - 11 6079 77
GenBank mRNA AF205066.1 - 7 - 8 791 77

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Recommended Assay Gallus gallus Probe spans exons

Panther Classification:

Molecular Function -

ligand-gated ion channel

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-vnb2r:80/100.66.79.197:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0