This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Gg03348710_m1
See other LPL GE Assays ›
Gene Symbol:
LPL
Gene Name:
lipoprotein lipase
Gene Aliases:
-
Chromosome Location:
Chr.Z: 54612194 - 54628984 on Build GRCg6a
Species:
Chicken
Species Specific ID (Flybase ID):
-
Assay ID Gg03348710_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LPL
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_205282.1 NP_990613.1 3 - 4 546 67
XM_015280414.2 XP_015135900.1 3 - 4 644 67
GenBank mRNA AB016987.1 - 3 - 4 519 67
EU477529.1 - 3 - 4 432 67
JX090309.1 - 3 - 4 544 67
X14670.1 - 546 67

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Gallus gallus Probe spans exons

Panther Classification:

Molecular Function -

lipase

Gene Ontology Categories:

Function(s) Process(es)

fatty acid metabolic process
fatty acid biosynthetic process
response to bacterium
positive regulation of macrophage derived foam cell differentiation
positive regulation of cholesterol storage
positive regulation of sequestering of triglyceride
lipid catabolic process
triglyceride catabolic process
cellular response to nutrient
chylomicron remodeling
very-low-density lipoprotein particle remodeling
cholesterol homeostasis
positive regulation of fat cell differentiation
positive regulation of inflammatory response
low-density lipoprotein particle mediated signaling
triglyceride homeostasis
cellular response to fatty acid
positive regulation of chemokine secretion
positive regulation of tumor necrosis factor secretion
lipoprotein lipase activity
phospholipase activity
triglyceride lipase activity
receptor binding
calcium ion binding
heparin binding
phosphatidylcholine 1-acylhydrolase activity
lipase activity
apolipoprotein binding
protein homodimerization activity
heparan sulfate proteoglycan binding
phosphatidylserine 1-acylhydrolase activity
1-acyl-2-lysophosphatidylserine acylhydrolase activity
lipoprotein particle binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-9hc65:80/100.66.78.230:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0