This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Gg03348795_gH
See other H3F3B GE Assays ›
Gene Symbol:
H3F3B
Gene Name:
H3 histone, family 3B
Gene Aliases:
H3-IX, H3-X, H3.3B
Chromosome Location:
Chr.18: 4707392 - 4709115 on Build GRCg6a
Species:
Chicken
Species Specific ID (Flybase ID):
-
Assay ID Gg03348795_gH
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: H3F3B
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_205296.1 NP_990627.1 2 - 3 233 71
GenBank mRNA Y00392.1 - 233 71

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Gallus gallus Probe spans exons

Panther Classification:

Molecular Function -

histone

Gene Ontology Categories:

Function(s) Process(es)

osteoblast differentiation
nucleus organization
spermatid development
single fertilization
cell proliferation
male gonad development
pericentric heterochromatin assembly
telomeric heterochromatin assembly
multicellular organism growth
muscle cell differentiation
oogenesis
regulation of centromere complex assembly
negative regulation of chromosome condensation
DNA binding
protein heterodimerization activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-ng27p:80/100.66.78.145:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline