This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Gg03349092_m1
See other GOT1 GE Assays ›
Gene Symbol:
GOT1
Gene Name:
glutamic-oxaloacetic transaminase 1
Gene Aliases:
cAspAT, cCAT
Chromosome Location:
Chr.6: 23099932 - 23103502 on Build GRCg6a
Species:
Chicken
Species Specific ID (Flybase ID):
-
Assay ID Gg03349092_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GOT1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_205321.1 NP_990652.1 5 - 6 662 93
GenBank mRNA X15636.1 - 662 93

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Recommended Assay Gallus gallus Probe spans exons

Panther Classification:

Molecular Function -

transaminase

Gene Ontology Categories:

Function(s) Process(es)

gluconeogenesis
2-oxoglutarate metabolic process
glycerol biosynthetic process
cellular amino acid metabolic process
aspartate metabolic process
aspartate biosynthetic process
glutamate metabolic process
L-aspartate:2-oxoglutarate aminotransferase activity
pyridoxal phosphate binding
L-cysteine:2-oxoglutarate aminotransferase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8cb8z:80/100.66.72.77:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0