This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Gg03349125_m1
See other RARB GE Assays ›
Gene Symbol:
RARB
Gene Name:
retinoic acid receptor beta
Gene Aliases:
RARBETA
Chromosome Location:
Chr.2: 37462374 - 37784739 on Build GRCg6a
Species:
Chicken
Species Specific ID (Flybase ID):
-
Assay ID Gg03349125_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: RARB
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_205326.1 NP_990657.1 8 - 9 1457 67
XM_015281313.2 XP_015136799.1 8 - 9 1635 67
XM_015281314.2 XP_015136800.1 8 - 9 1469 67
XM_015281315.2 XP_015136801.1 8 - 9 1488 67
XM_015281316.2 XP_015136802.1 6 - 7 2163 67
XM_015281317.2 XP_015136803.1 6 - 7 1166 67
XM_025147066.1 XP_025002834.1 7 - 8 1355 67
XM_025147067.1 XP_025002835.1 7 - 8 1305 67
GenBank mRNA S63196.1 - 6 - 7 924 67
X56674.1 - 1309 67
X57339.1 - 1132 67
X57340.1 - 1113 67
X57341.1 - 1457 67
X59473.1 - 1001 67

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Gallus gallus Probe spans exons

Panther Classification:

Molecular Function -

C4 zinc finger nuclear receptor

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
ureteric bud development
glandular epithelial cell development
outflow tract septum morphogenesis
growth plate cartilage development
negative regulation of cell proliferation
hormone-mediated signaling pathway
striatum development
cell differentiation
negative regulation of chondrocyte differentiation
response to retinoic acid
limb morphogenesis
embryonic hindlimb morphogenesis
multicellular organism growth
positive regulation of apoptotic process
negative regulation of apoptotic process
positive regulation of transcription from RNA polymerase II promoter
embryonic eye morphogenesis
retinoic acid receptor signaling pathway
embryonic digestive tract development
ventricular cardiac muscle cell differentiation
inner ear receptor cell differentiation involved in inner ear sensory epithelium regeneration
beak morphogenesis
RNA polymerase II core promoter proximal region sequence-specific DNA binding
RNA polymerase II transcription factor activity, ligand-activated sequence-specific DNA binding
zinc ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l2rv2:80/100.66.78.26:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0