This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Gg03350418_m1
See other TWF2 GE Assays ›
Gene Symbol:
TWF2
Gene Name:
twinfilin actin binding protein 2
Gene Aliases:
PTK9L
Chromosome Location:
Chr.12: 3353840 - 3372303 on Build GRCg6a
Species:
Chicken
Species Specific ID (Flybase ID):
-
Assay ID Gg03350418_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: TWF2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001030589.1 NP_001025760.1 3 - 4 364 62
XM_015292829.1 XP_015148315.1 3 - 4 382 62
XM_015292830.1 XP_015148316.1 3 - 4 382 62
XM_015292831.2 XP_015148317.1 3 - 4 395 62
XM_015292832.1 XP_015148318.1 3 - 4 382 62
XM_015292833.1 XP_015148319.1 3 - 4 382 62
XM_015292834.1 XP_015148320.1 3 - 4 382 62
XM_015292835.1 XP_015148321.1 3 - 4 382 62
XM_015292836.1 XP_015148322.1 3 - 4 382 62
XM_015292837.2 XP_015148323.1 3 - 4 397 62
GenBank mRNA AJ719549.1 - 3 - 4 364 62

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Gallus gallus Probe spans exons

Panther Classification:

Molecular Function -

non-motor actin binding protein

Gene Ontology Categories:

Function(s) Process(es)

regulation of lamellipodium assembly
positive regulation of neuron projection development
actin filament depolymerization
sequestering of actin monomers
barbed-end actin filament capping
actin monomer binding
actin filament binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2k58s:80/100.66.76.192:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0