This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Gg03351780_m1
See other LCLAT1 GE Assays ›
Gene Symbol:
LCLAT1
Gene Name:
lysocardiolipin acyltransferase 1
Gene Aliases:
1-AGPAT 8, LYCAT
Chromosome Location:
Chr.3: 8530367 - 8637204 on Build GRCg6a
Species:
Chicken
Species Specific ID (Flybase ID):
-
Assay ID Gg03351780_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LCLAT1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001031039.1 NP_001026210.1 2 - 3 298 72
XM_015278082.2 XP_015133568.1 3 - 4 510 72
XM_015278083.2 XP_015133569.1 3 - 4 366 72
XM_015278084.2 XP_015133570.1 3 - 4 476 72
XM_015278085.2 XP_015133571.1 2 - 3 300 72
XM_025148656.1 XP_025004424.1 4 - 5 556 72
XM_025148657.1 XP_025004425.1 2 - 3 327 72
XM_025148658.1 XP_025004426.1 4 - 5 556 72
GenBank mRNA AJ851530.1 - 2 - 3 298 72

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Recommended Assay Gallus gallus Probe spans exons

Panther Classification:

Molecular Function -

acyltransferase

Gene Ontology Categories:

Function(s) Process(es)

multicellular organism development
CDP-diacylglycerol biosynthetic process
phosphatidylinositol acyl-chain remodeling
1-acylglycerol-3-phosphate O-acyltransferase activity
transferase activity, transferring acyl groups

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mvcqg:80/100.66.75.199:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0