This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Gg03352049_m1
See other ASXL2 GE Assays ›
Gene Symbol:
ASXL2
Gene Name:
additional sex combs like 2, transcriptional regulator
Gene Aliases:
-
Chromosome Location:
Chr.3: 104744834 - 104834408 on Build GRCg6a
Species:
Chicken
Species Specific ID (Flybase ID):
-
Assay ID Gg03352049_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ASXL2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001031096.1 NP_001026267.1 7 - 8 952 66
XM_015285098.2 XP_015140584.1 7 - 8 960 66
XM_015285099.2 XP_015140585.1 9 - 10 851 66
XM_015285100.2 XP_015140586.1 4 - 5 545 66
XM_025148790.1 XP_025004558.1 7 - 8 798 66
XM_025148791.1 XP_025004559.1 7 - 8 677 66
XM_025148792.1 XP_025004560.1 6 - 7 833 66
GenBank mRNA AJ719496.1 - 7 - 8 952 66

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Gallus gallus Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
organ morphogenesis
positive regulation of transcription from RNA polymerase II promoter
DNA binding
chromatin binding
peroxisome proliferator activated receptor binding
metal ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2xklz:80/100.66.76.139:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0