This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Gg03352725_g1
See other SEC22B GE Assays ›
Gene Symbol:
SEC22B
Gene Name:
SEC22 homolog B, vesicle trafficking protein
Gene Aliases:
SEC22L1
Chromosome Location:
Chr.8: 4531053 - 4539913 on Build GRCg6a
Species:
Chicken
Species Specific ID (Flybase ID):
-
Assay ID Gg03352725_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SEC22B
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001031266.3 NP_001026437.1 1 - 2 137 65
XM_025152808.1 XP_025008576.1 1 - 2 134 65
GenBank mRNA AJ720320.1 - 1 - 2 112 65
BU255647.1 - 128 65

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Gallus gallus Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

ER to Golgi vesicle-mediated transport
retrograde vesicle-mediated transport, Golgi to ER
protein transport
positive regulation of protein catabolic process
vesicle fusion with Golgi apparatus
negative regulation of autophagosome assembly
SNAP receptor activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-vnb2r:80/100.66.79.197:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0