This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Gg03353626_m1
See other PCMT1 GE Assays ›
Gene Symbol:
PCMT1
Gene Name:
protein-L-isoaspartate (D-aspartate) O-methyltransferase
Gene Aliases:
-
Chromosome Location:
Chr.3: 48319791 - 48353946 on Build GRCg6a
Species:
Chicken
Species Specific ID (Flybase ID):
-
Assay ID Gg03353626_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PCMT1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001031525.1 NP_001026696.1 6 - 7 556 61
XM_015284116.2 XP_015139602.2 6 - 7 595 61
GenBank mRNA AJ851619.1 - 6 - 7 556 61

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Recommended Assay Gallus gallus Probe spans exons

Panther Classification:

Molecular Function -

methyltransferase

Gene Ontology Categories:

Function(s) Process(es)

protein methylation
protein-L-isoaspartate (D-aspartate) O-methyltransferase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-ng27p:80/100.66.78.145:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline