This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Gg03354355_m1
See other ORAI1 GE Assays ›
Gene Symbol:
ORAI1
Gene Name:
ORAI calcium release-activated calcium modulator 1
Gene Aliases:
TMEM142A, orai-1
Chromosome Location:
Chr.15: 5872047 - 5889252 on Build GRCg6a
Species:
Chicken
Species Specific ID (Flybase ID):
-
Assay ID Gg03354355_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ORAI1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001030658.1 NP_001025829.1 1 - 2 290 98
GenBank mRNA AJ719929.1 - 1 - 2 290 98

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 3' Most Recommended Assay Gallus gallus Probe spans exons

Panther Classification:

Molecular Function -

ion channel transporter

Gene Ontology Categories:

Function(s) Process(es)

store-operated calcium entry
adaptive immune response
positive regulation of calcium ion transport
calcium ion transmembrane transport
store-operated calcium channel activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-lzcpb:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0