This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Gg03354546_m1
See other GTF3C5 GE Assays ›
Gene Symbol:
GTF3C5
Gene Name:
general transcription factor IIIC subunit 5
Gene Aliases:
-
Chromosome Location:
Chr.17: 7080377 - 7089081 on Build GRCg6a
Species:
Chicken
Species Specific ID (Flybase ID):
-
Assay ID Gg03354546_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GTF3C5
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001030684.1 NP_001025855.1 4 - 5 771 69
XM_025156063.1 XP_025011831.1 4 - 5 778 69
XM_025156064.1 XP_025011832.1 4 - 5 778 69
GenBank mRNA AJ719580.1 - 4 - 5 771 69

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Gallus gallus Probe spans exons

Panther Classification:

Molecular Function -

RNA metabolism protein nucleic acid metabolism protein general transcription factor

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rpjph:80/100.66.75.107:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0