This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Gg03354751_m1
See other ORAI3 GE Assays ›
Gene Symbol:
ORAI3
Gene Name:
ORAI calcium release-activated calcium modulator 3
Gene Aliases:
ORAI2
Chromosome Location:
Chr.19: 4284069 - 4296320 on Build GRCg6a
Species:
Chicken
Species Specific ID (Flybase ID):
-
Assay ID Gg03354751_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ORAI3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001030710.1 NP_001025881.1 3 - 4 373 85
XM_025141935.1 XP_024997703.1 5 - 6 2351 85
XM_025141936.1 XP_024997704.1 5 - 6 3092 85
XM_025141937.1 XP_024997705.1 5 - 6 2892 85
XM_025141938.1 XP_024997706.1 3 - 4 3147 85
XM_025141939.1 XP_024997707.1 3 - 4 2689 85
GenBank mRNA AJ719622.1 - 3 - 4 373 85

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Recommended Assay Gallus gallus Probe spans exons

Panther Classification:

Molecular Function -

ion channel transporter

Gene Ontology Categories:

Function(s) Process(es)

store-operated calcium entry
calcium ion transmembrane transport
store-operated calcium channel activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-f5rhb:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0