This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Gg03354986_m1
See other TWF1 GE Assays ›
Gene Symbol:
TWF1
Gene Name:
twinfilin actin binding protein 1
Gene Aliases:
PTK9
Chromosome Location:
Chr.1: 30203889 - 30222348 on Build GRCg6a
Species:
Chicken
Species Specific ID (Flybase ID):
-
Assay ID Gg03354986_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: TWF1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001030739.1 NP_001025910.1 3 - 4 364 128
NM_001278095.1 NP_001265024.1 3 - 4 364 128
GenBank mRNA AJ719701.1 - 3 - 4 364 128
BU464074.1 - 3 - 4 308 128

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Recommended Assay Gallus gallus Probe spans exons

Panther Classification:

Molecular Function -

non-motor actin binding protein

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-w8bxd:80/100.66.79.51:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0