This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Gg03355315_m1
See other CD200R1L GE Assays ›
Gene Symbol:
CD200R1L
Gene Name:
CD200 receptor 1-like
Gene Aliases:
CD200R-S1, CD200R1B
Chromosome Location:
Chr.1: 84533618 - 84555217 on Build GRCg6a
Species:
Chicken
Species Specific ID (Flybase ID):
-
Assay ID Gg03355315_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CD200R1L
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001030782.2 NP_001025953.2 5 - 6 869 130
GenBank mRNA AJ720790.1 - 5 - 6 869 130

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons

Panther Classification:

Molecular Function -

transmembrane signal receptor

Gene Ontology Categories:

Function(s) Process(es)

regulation of neuroinflammatory response
signaling receptor activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-wt24k:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline