This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Gg03355925_m1
See other GPAT3 GE Assays ›
Gene Symbol:
GPAT3
Gene Name:
glycerol-3-phosphate acyltransferase 3
Gene Aliases:
AGPAT 10, AGPAT9, LPAAT-THETA
Chromosome Location:
Chr.4: 46506402 - 46524034 on Build GRCg6a
Species:
Chicken
Species Specific ID (Flybase ID):
-
Assay ID Gg03355925_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GPAT3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001031145.1 NP_001026316.1 2 - 3 294 70
XM_015276269.1 XP_015131755.1 2 - 3 180 70
XM_015276270.1 XP_015131756.1 2 - 3 200 70
GenBank mRNA AJ719716.1 - 2 - 3 294 70

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Gallus gallus Probe spans exons

Panther Classification:

Molecular Function -

acyltransferase

Gene Ontology Categories:

Function(s) Process(es)

CDP-diacylglycerol biosynthetic process
triglyceride biosynthetic process
1-acylglycerol-3-phosphate O-acyltransferase activity
glycerol-3-phosphate O-acyltransferase activity
sn-1-glycerol-3-phosphate C16:0-DCA-CoA acyl transferase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-d7bxm:80/100.66.76.204:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0