This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Gg03356283_m1
See other MIER1 GE Assays ›
Gene Symbol:
MIER1
Gene Name:
MIER1 transcriptional regulator
Gene Aliases:
-
Chromosome Location:
Chr.8: 28936348 - 28967729 on Build GRCg6a
Species:
Chicken
Species Specific ID (Flybase ID):
-
Assay ID Gg03356283_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MIER1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001031292.1 NP_001026463.1 11 - 12 1105 73
XM_015291033.2 XP_015146519.1 11 - 12 1191 73
XM_015291034.2 XP_015146520.1 11 - 12 1172 73
XM_015291035.2 XP_015146521.1 11 - 12 1229 73
XM_015291036.2 XP_015146522.1 12 - 13 1296 73
XM_015291037.2 XP_015146523.1 9 - 10 1065 73
XM_025152887.1 XP_025008655.1 10 - 11 1062 73
GenBank mRNA AJ719995.1 - 11 - 12 1105 73
CR407018.1 - 165 73

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Recommended Assay Gallus gallus Probe spans exons

Panther Classification:

Molecular Function -

homeodomain transcription factor

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
histone deacetylation
positive regulation of chromatin silencing
RNA polymerase II repressing transcription factor binding
transcription corepressor activity
histone deacetylase binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0