This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Gg03358987_m1
See other ALDOB GE Assays ›
Gene Symbol:
ALDOB
Gene Name:
aldolase, fructose-bisphosphate B
Gene Aliases:
ALDB
Chromosome Location:
Chr.Z: 65329391 - 65338114 on Build GRCg6a
Species:
Chicken
Species Specific ID (Flybase ID):
-
Assay ID Gg03358987_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ALDOB
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001007977.3 NP_001007978.1 5 - 6 673 60
GenBank mRNA CR523513.1 - 5 - 6 602 60

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Gallus gallus Probe spans exons

Panther Classification:

Molecular Function -

aldolase

Gene Ontology Categories:

Function(s) Process(es)

liver development
fructose metabolic process
gluconeogenesis
glycolytic process
NADH oxidation
fructose 1,6-bisphosphate metabolic process
positive regulation of ATPase activity
vacuolar proton-transporting V-type ATPase complex assembly
fructose-bisphosphate aldolase activity
cytoskeletal protein binding
identical protein binding
ATPase binding
fructose-1-phosphate aldolase activity
fructose binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0