This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Gg03360149_m1
See other PCDH1 GE Assays ›
Gene Symbol:
PCDH1
Gene Name:
protocadherin 1
Gene Aliases:
protocadherin-1
Chromosome Location:
Chr.13: 2787812 - 2837441 on Build GRCg6a
Species:
Chicken
Species Specific ID (Flybase ID):
-
Assay ID Gg03360149_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PCDH1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001045827.1 NP_001039292.1 1 - 2 63 102
XM_015293599.2 XP_015149085.1 1 - 2 342 102
XM_015293600.2 XP_015149086.1 1 - 2 339 102
GenBank mRNA AY676328.1 - 1 - 2 63 102

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Gallus gallus Probe spans exons

Panther Classification:

Molecular Function -

cadherin

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-fzgxv:80/100.66.76.103:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0