This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Gg03361113_m1
See other TNFRSF19 GE Assays ›
Gene Symbol:
TNFRSF19
Gene Name:
TNF receptor superfamily member 19
Gene Aliases:
-
Chromosome Location:
Chr.1: 178822907 - 178879732 on Build GRCg6a
Species:
Chicken
Species Specific ID (Flybase ID):
-
Assay ID Gg03361113_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: TNFRSF19
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001080103.1 NP_001073572.1 4 - 5 477 67
XM_015278030.2 XP_015133516.1 6 - 7 813 67
XM_015278040.2 XP_015133526.1 5 - 6 544 67
XM_025150569.1 XP_025006337.1 6 - 7 574 67
GenBank mRNA BX931334.2 - 4 - 5 480 67
BX933700.2 - 5 - 6 547 67
DQ360499.1 - 4 - 5 477 67
DQ360500.1 - 4 - 5 477 67

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Gallus gallus Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0