This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Gg03361851_m1
See other CDH8 GE Assays ›
Gene Symbol:
CDH8
Gene Name:
cadherin 8
Gene Aliases:
cadherin-8
Chromosome Location:
Chr.11: 12823945 - 12973998 on Build GRCg6a
Species:
Chicken
Species Specific ID (Flybase ID):
-
Assay ID Gg03361851_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CDH8
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001100289.1 NP_001093759.1 11 - 12 2425 95
XM_015292511.2 XP_015147997.2 12 - 13 2594 95
XM_015292512.2 XP_015147998.1 2 - 3 2278 95
XM_015292513.2 XP_015147999.1 3 - 4 514 95
XM_025154214.1 XP_025009982.1 3 - 4 498 95
XM_025154215.1 XP_025009983.1 2 - 3 379 95
GenBank mRNA EF608154.1 - 11 - 12 2425 95

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 3' Most Recommended Assay Gallus gallus Probe spans exons

Panther Classification:

Molecular Function -

cadherin

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-cjlz8:80/100.66.74.50:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0