This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Gg03362793_m1
See other BMF GE Assays ›
Gene Symbol:
BMF
Gene Name:
Bcl2 modifying factor
Gene Aliases:
-
Chromosome Location:
Chr.5: 29607647 - 29631286 on Build GRCg6a
Species:
Chicken
Species Specific ID (Flybase ID):
-
Assay ID Gg03362793_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: BMF
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001113292.1 NP_001106763.1 2 - 3 256 56
XM_015287560.1 XP_015143046.1 3 - 4 1115 56
XM_015287561.2 XP_015143047.1 4 - 5 744 56
XM_015287563.2 XP_015143049.1 4 - 5 1283 56
XM_015287564.2 XP_015143050.1 3 - 4 1119 56
XM_015287565.1 XP_015143051.1 3 - 4 890 56
XM_025150841.1 XP_025006609.1 4 - 5 1055 56
XM_025150842.1 XP_025006610.1 3 - 4 899 56
GenBank mRNA EF621525.1 - 2 - 3 481 56
EF621526.1 - 2 - 3 256 56

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Recommended Assay Gallus gallus Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0