This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Gg03362824_m1
See other GTF3A GE Assays ›
Gene Symbol:
GTF3A
Gene Name:
general transcription factor IIIA
Gene Aliases:
-
Chromosome Location:
Chr.1: 177594912 - 177599321 on Build GRCg6a
Species:
Chicken
Species Specific ID (Flybase ID):
-
Assay ID Gg03362824_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GTF3A
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001113571.1 NP_001107043.1 5 - 6 644 100
GenBank mRNA BU225193.1 - 1 - 2 97 100
BU349430.1 - 2 - 3 97 100
CR352757.1 - 5 - 6 647 100
CR406086.1 - 3 - 4 216 100

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Recommended Assay Gallus gallus Probe spans exons

Panther Classification:

Molecular Function -

C2H2 zinc finger transcription factor

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-9dqhd:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline