This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Gg03363421_m1
See other CDH20 GE Assays ›
Gene Symbol:
CDH20
Gene Name:
cadherin 20, type 2
Gene Aliases:
-
Chromosome Location:
Chr.2: 68583727 - 68927139 on Build GRCg6a
Species:
Chicken
Species Specific ID (Flybase ID):
-
Assay ID Gg03363421_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CDH20
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_204134.1 NP_989465.1 9 - 10 1715 67
XM_015282158.2 XP_015137644.1 10 - 11 1865 67
XM_015282159.2 XP_015137645.1 9 - 10 1735 67
XM_015282160.2 XP_015137646.1 9 - 10 1714 67
XM_015282161.2 XP_015137647.1 8 - 9 1780 67
XM_025146992.1 XP_025002760.1 9 - 10 1854 67
GenBank mRNA AF459439.1 - 9 - 10 1715 67
AF465257.1 - 9 - 10 1740 67
AF465258.1 - 3 - 4 978 67

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Gallus gallus Probe spans exons

Panther Classification:

Molecular Function -

cadherin

Gene Ontology Categories:

Function(s) Process(es)

cell morphogenesis
cell-cell junction assembly
homophilic cell adhesion via plasma membrane adhesion molecules
multicellular organism development
calcium-dependent cell-cell adhesion via plasma membrane cell adhesion molecules
adherens junction organization
cell-cell adhesion via plasma-membrane adhesion molecules
calcium ion binding
cadherin binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8rxhs:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0