This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Gg03363688_m1
See other BRCA1 GE Assays ›
Gene Symbol:
BRCA1
Gene Name:
BRCA1 DNA repair associated
Gene Aliases:
-
Chromosome Location:
Chr.27: 7969221 - 7990488 on Build GRCg6a
Species:
Chicken
Species Specific ID (Flybase ID):
-
Assay ID Gg03363688_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: BRCA1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_204169.1 NP_989500.1 21 - 22 5253 63
XM_015299561.2 XP_015155047.1 20 - 21 5241 63
XM_015299562.2 XP_015155048.1 21 - 22 5250 63
XM_015299563.2 XP_015155049.1 19 - 20 5160 63
XM_015299565.2 XP_015155051.1 17 - 18 5112 63
XM_025143941.1 XP_024999709.1 20 - 21 5238 63
GenBank mRNA AF355273.1 - 21 - 22 5253 63

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Recommended Assay Gallus gallus Probe spans exons

Panther Classification:

Molecular Function -

ubiquitin-protein ligase protein modifying enzyme

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2k58s:80/100.66.76.192:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0