This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Gg03363801_m1
See other CDH7 GE Assays ›
Gene Symbol:
CDH7
Gene Name:
cadherin 7
Gene Aliases:
cadherin-7
Chromosome Location:
Chr.2: 95468521 - 95545982 on Build GRCg6a
Species:
Chicken
Species Specific ID (Flybase ID):
-
Assay ID Gg03363801_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CDH7
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_204187.2 NP_989518.2 7 - 8 1723 67
GenBank mRNA AY115555.1 - 7 - 8 1723 67
D42150.1 - 7 - 8 1723 67

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Gallus gallus Probe spans exons

Panther Classification:

Molecular Function -

cadherin

Gene Ontology Categories:

Function(s) Process(es)

cell morphogenesis
cartilage condensation
neural tube formation
cell-cell junction assembly
homophilic cell adhesion via plasma membrane adhesion molecules
multicellular organism development
calcium-dependent cell-cell adhesion via plasma membrane cell adhesion molecules
cell migration
adherens junction organization
neural crest cell delamination
wound healing
limb development
chondroblast differentiation
cell-cell adhesion via plasma-membrane adhesion molecules
calcium ion binding
cadherin binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-zvjvc:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline